• USA Home
  • F9924 - Fibroblast Growth Factor Receptor-3α (IIIc)/Fc Chimera from mouse

EMAIL THIS PAGE TO A FRIEND
F9924 Sigma

Fibroblast Growth Factor Receptor-3α (IIIc)/Fc Chimera from mouse

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: FGFR-3α (IIIc)/Fc Chimera

Purchase

Description

Analysis Note

The bioactivity is measured by its ability to inhibit human fibroblast growth factor acidic-dependent proliferation of NR6 cells

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?