• USA Home
  • FFT15 - Firefly tails, desiccated, preweighed vials

EMAIL THIS PAGE TO A FRIEND
FFT15 Sigma

Firefly tails, desiccated, preweighed vials

Purchase

Properties

Related Categories Biochemicals and Reagents, Protein Extracts, Proteins and Derivatives More...
usage   sufficient for 1 demonstration (of bioluminescence)
packaging   vial of 15 mg powdered, desiccated firefly tails
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?