• USA Home
  • G0131 - Glyoxalase II from bovine liver

EMAIL THIS PAGE TO A FRIEND
G0131 Sigma

Glyoxalase II from bovine liver

lyophilized powder, ~10 units/mg protein

Synonym: S-2-Hydroxyacylglutathione hydrolase

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   lyophilized powder
composition   Protein, ~80% modified Warburg-Christian
foreign activity   glyoxalase I <0.2%
storage temp.   −20°C

Description

Components

Lyophilized powder containing approximately 80% protein (modified Warburg-Christian); balance primarily phosphate-citrate buffer salts.

Reconstitution

Reconstitute with H2O to 5-10 mg protein/ml, and store frozen. If lower condentration is desired, dilute with 5 mM KH2PO4, 0.1% BSA, pH 7.4.

Unit Definition

One unit will hydrolyze 1.0 μmole of S-lactoylglutathione per min at pH 7.4 at 25°C.

Physical form

Lyophilized powder containing phosphate-citrate buffer salts

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?