• USA Home
  • G0139 - L-α-Glycerophospho-D-myo-inositol 4-monophosphate Potassium salt from Bovine Brain L-α-phosphatidylinositol 4-monophosphate

EMAIL THIS PAGE TO A FRIEND
G0139 Sigma

L-α-Glycerophospho-D-myo-inositol 4-monophosphate Potassium salt from Bovine Brain L-α-phosphatidylinositol 4-monophosphate

0.5 mg/mL in ethanol: water (3:1), ~98% (TLC)

Purchase

Properties

assay   ~98% (TLC)
concentration   0.5 mg/mL in ethanol: water (3:1)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?