EMAIL THIS PAGE TO A FRIEND
G0254 Sigma

Gly-Gly-Ala

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Peptides, Tripeptides
InChI Key   CCQOOWAONKGYKQ-BYPYZUCNSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?