• USA Home
  • G0261 - γ-Globulins from rabbit

EMAIL THIS PAGE TO A FRIEND
G0261 Sigma

γ-Globulins from rabbit

99% (electrophoresis)

Purchase

Properties

assay   99% (electrophoresis)
impurities   <5% NaCl
storage temp.   −20°C

Description

Preparation Note

Prepared from Cohn Fraction II, III

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

G2018 γ-Globulins from rabbit, ≥99% (agarose gel electrophoresis)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?