• USA Home
  • G0273 - Grace’s amino acids solution

EMAIL THIS PAGE TO A FRIEND
G0273 Sigma

Grace’s amino acids solution

10 ×, Without L-glutamine, sterile-filtered, insect cell culture tested

Purchase

Properties

sterility   sterile-filtered
concentration   10 ×
impurities   endotoxin, tested
suitability   insect cell culture tested
storage temp.   2-8°C

Description

General description

This is a 10X stock solution of Grace′s Amino Acids without Glutamine.

Grace′s medium was originally formulated to support the growth of cells derived from the Australian emperor gum moth, Antherea eucalypti. The medium is a modification of Wyatt′s medium to more closely resemble Antherea hemolymph. The unparalled success achieved by Grace in maintaining long term cultures of a variety of insect cells led to a resurgence in the field of insect tissue culture and wide usage of the medium. Many modifications of this medium have been developed since its introduction.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?