• USA Home
  • G0389 - Glyceraldehyde-3-phosphate Dehydrogenase Agarose from baker's yeast (S. cerevisiae)

EMAIL THIS PAGE TO A FRIEND
G0389 Sigma

Glyceraldehyde-3-phosphate Dehydrogenase Agarose from baker's yeast (S. cerevisiae)

lyophilized powder, 50-150 units/g dry agarose, pH 7.6, 30 °C

Synonym: D-Glyceraldehyde 3-phosphate: NAD+ oxidoreductase (phosphorylating), GAPDH

Purchase

Description

Biochem/physiol Actions

GAPDH catalyzes the conversion of glyceraldehyde 3-phosphate to glycerate 1,3-biphosphate. GAPDH is essential for both glycolysis and gluconeogenesis.

Quality

One mL gel will yield 1-5 units.

Unit Definition

One unit will reduce 1.0 μmole of 3-phosphoglycerate to D-glyceraldehyde 3-phosphate per min in a coupled system with 3-phosphoglyceric phosphokinase at pH 7.6 at 25°C, unless otherwise indicated.

Physical form

Lyophilized powder, stabilized with lactose, citrate and cysteine

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Articles

Insulin Signaling and Energy Homeostasis

Glucose metabolism is regulated by the opposing actions of insulin and glucagon. Insulin is released from pancreatic ß cells in response to high blood glucose levels and regulates glucose metabolism ...
Linda Stephenson, Ph.D.
Biofiles v6 n4, 2011
Keywords: Apoptosis, Biological processes, Carboxylations, Catalog, Diabetes, Gene expression, Gluconeogenesis, Glycolysis, Hormones, Metabolism, Metabolites, Transcription, Transduction, Type

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

inhibitor

Product #

Description

Add to Cart

I2512 Sodium iodoacetate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?