• USA Home
  • G0400 - L-Glutamate Oxidase from Streptomyces sp.

EMAIL THIS PAGE TO A FRIEND
G0400 Sigma

L-Glutamate Oxidase from Streptomyces sp.

lyophilized powder, ≥5.0 units/mg solid

Synonym: L-Glutamate:oxygen oxidoreductase (deaminating)

Purchase

Description

Biochem/physiol Actions

Major enzyme in the synthesis or degradation of glutamic acid; transfers an amine to α-ketoglutaric acid to form L-glutamic acid or deamidates L-glutamic acid

Unit Definition

One unit will form 1.0 μmole of α-ketoglutaric acid from L-glutamic acid per min at pH 7.4 at 30 °C.

Physical form

Lyophilized powder containing potassium phosphate buffer salts and lactose

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?