• USA Home
  • G0421 - Poly-L-glutamic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
G0421 Sigma

Poly-L-glutamic acid sodium salt

mol wt >50,000

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, Homopolymers, Polyamino Acids
mol wt   mol wt >50,000
storage temp.   −20°C

Description

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Biochem/physiol Actions

Poly-L-glutamic acid is an anionic biocompatible polymer used in combination with cationic biopolymers such as poly-lysine to form stable time-release controlled polyelectrolyte shells to protect drug and molecular delivery structures such as protein-loaded starch microgels and nanocapsules.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Poly(Amino Acid) Block Copolymers for Drug Delivery and other Biomedical Applications

Carmen Scholz Department of Chemistry, University of Alabama in Huntsville Huntsville, AL 35899 Email: scholzc@uah.edu
Keywords: Addition reactions, Asymmetric synthesis, Building blocks, Catalysis, Degradations, Living polymerization, PEGylations, Polymerization reactions, Precipitation, Proteomics, Ring-opening polymerization, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?