• USA Home
  • G0504 - D-Gulonic acid γ-lactone

EMAIL THIS PAGE TO A FRIEND
G0504 Sigma

D-Gulonic acid γ-lactone

Synonym: D(−)-Gulono-1,4-lactone

  • CAS Number 6322-07-2

  • Empirical Formula (Hill Notation) C6H10O6

  • Molecular Weight 178.14

  •  Beilstein Registry Number 83012

  •  EC Number 228-678-5

  •  MDL number MFCD00005391

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   SXZYCXMUPBBULW-LECHCGJUSA-N

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

219355 D-Gulonic acid γ-lactone, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?