• USA Home
  • G0649 - Glucose Dehydrogenase, NADP dependent from Cryptococcus uniguttulatus

EMAIL THIS PAGE TO A FRIEND
G0649 Sigma

Glucose Dehydrogenase, NADP dependent from Cryptococcus uniguttulatus

lyophilized powder, ≥10 units/mg solid

Synonym: β-D-Glucose: NADP+ 1-oxidoreductase

Purchase

Description

Unit Definition

One unit will oxidize 1.0 μmole of β-D-glucose to D-glucono-δ-lactone per min at pH 8.0 at 37 °C in the presence of NADP+.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?