• USA Home
  • G0653 - Glyceraldehyde-3-phosphate Dehydrogenase, Carbamylated 2D Electrophoresis Marker from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
G0653 Sigma

Glyceraldehyde-3-phosphate Dehydrogenase, Carbamylated 2D Electrophoresis Marker from rabbit muscle

Purchase

Properties

Related Categories 2D Markers, Isoelectric Focusing and Two Dimensional Electrophoresis, Molecular Biology, Protein Electrophoresis, Proteomics More...
mol wt   mol wt ~36,000
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?