• USA Home
  • G0763 - Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
G0763 Sigma

Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle

ammonium sulfate suspension, 80-120 units/mg protein (biuret)

Synonym: D-Glyceraldehyde 3-phosphate:NAD+ oxidoreductase (phosphorylating), GAPDH

Purchase

Properties

Related Categories 1.2.x.x Acting on aldehydes or oxo groups, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   ammonium sulfate suspension
foreign activity   3-Phosphoglyceric phosphokinase <0.05%
  triosephosphate isomerase, lactic dehydrogenase, myokinase, and pyruvate kinase <0.01%
storage temp.   2-8°C

Description

Unit Definition

One unit will reduce 1.0 μmole of 3-phosphoglycerate to D-glyceraldehyde 3-phosphate per min in a coupled system with 3-phosphoglyceric phosphokinase at pH 7.6 at 25 °C.

Physical form

Crystalline suspension in 2.6 M (NH4)2SO4, pH 7.5, containing 1 mM EDTA and 0.02 M 2-mercaptoethanol

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase catalyzes the conversion of glyceraldehyde-3-phosphate into D-glycerate-1,3-bisphosphate as part of the glycolysis pathway.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

G2267 Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein

inhibitor

Product #

Description

Add to Cart

I2512 Sodium iodoacetate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?