EMAIL THIS PAGE TO A FRIEND
G0767 Sigma

Gly-Phe-Ser

  • CAS Number 23828-14-0

  • Empirical Formula (Hill Notation) C14H19N3O5

  • Molecular Weight 309.32

  •  MDL number MFCD00058472

  •  PubChem Substance ID 24895024

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Peptides, Tripeptides
InChI Key   GGAPHLIUUTVYMX-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?