G0918 Sigma
Gel Mount Aqueous Mounting Medium
DOWNLOAD MSDS (PDF)
Description
Application
Permanent aqueous mounting medium optimized for preservation of fluorescent-stained tissue sections. Suitable for use with conjugated tracers including fluorscein, rhodamine, Texas Red®, phycoerythrin, and phycocyanin. Gel Mount® is a trademark of Biomeda Corp.
Legal Information
3M is a registered trademark of 3M Company
Gel Mount is a trademark of Biomeda Corp.
Texas Red is a registered trademark of Life Technologies Corporation
Price and Availability
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?