EMAIL THIS PAGE TO A FRIEND
G106 Sigma

Glibenclamide

Synonym: 5-Chloro-N-(2-[4-([(cyclohexylamino]carbonyl)amino]sulfonyl)phenyl]ethyl)-2-methoxybenzamide, Glyburide

Purchase

Properties

form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2
RTECS 
YS4725200

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G0639 Glybenclamide, ≥99% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?