• USA Home
  • G1141 - Gly-Gly benzyl ester p-toluenesulfonate salt

EMAIL THIS PAGE TO A FRIEND
G1141 Sigma

Gly-Gly benzyl ester p-toluenesulfonate salt

  • CAS Number 1738-82-5

  • Empirical Formula (Hill Notation) C11H14N2O3 · C7H8O3S

  • Molecular Weight 394.44

  •  MDL number MFCD00050678

  •  PubChem Substance ID 24895043

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   RWBJLSORGVEMHA-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Amino Acid Sequence

Gly-Gly-OBz

Application

Gly-Gly benzyl ester p-toluenesulfonate salt may be used as an organic intermediate in peptide synthetic processes.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?