EMAIL THIS PAGE TO A FRIEND
G120 Sigma

GYKI 52895

solid

Synonym: 1-(4-Aminophenyl)-4-methyl-7,8-methylenedioxy-3,4-dihydro-5H-2,3-benzodiazepine

  • Empirical Formula (Hill Notation) C17H17N3O2

  • Molecular Weight 295.34

  •  MDL number MFCD00153804

  •  PubChem Substance ID 24277937

Purchase

Properties

Related Categories Biochemicals and Reagents, Biogenic Amine Transport Inhibitors, Cell Biology, Cell Signaling and Neuroscience, GPCR Modulators,
InChI Key   AQTITSBNGSVQNZ-UHFFFAOYSA-N
form   solid
color   white
solubility   ethanol: soluble
Gene Information   human ... DBH(1621)
rat ... Gabra2(29706), Gria1(50592), Grik1(29559), Grin2a(24409)

Description

Biochem/physiol Actions

Selective dopamine uptake inhibitor; antiparkinsonian; antidepressant.

Legal Information

Sold under exclusive license from the Institute for Drug Research Ltd. Budapest, Hungary.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?