• USA Home
  • G1273 - Rollpruf sheer natural latex rubber gloves
EMAIL THIS PAGE TO A FRIEND
G1273 Sigma
Rollpruf sheer natural latex rubber gloves
size 7
Purchase
Properties
size   7
availability   not available in EU
L × thickness   11 in. × 0.008 in.
Description
General description
Surgical quality natural latex has no extractable plasticizer and is powder-free.
Final rinse with filtered deionized water reduces ion content. Cuff band is color-coded by size; band prevents rolldown and secures gloves on shirt sleeves. Finished, inspected, tested, and packaged in a Class100 clean room.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?