• USA Home
  • G1274 - HA-1004 hydrochloride

EMAIL THIS PAGE TO A FRIEND
G1274 Sigma

HA-1004 hydrochloride

≥98% (HPLC), powder

Synonym: N-(2-Guanidinoethyl)-5-isoquinolinesulfonamide hydrochloride

  • CAS Number 92564-34-6

  • Empirical Formula (Hill Notation) C12H15N5O2S · HCl

  • Molecular Weight 329.81

  •  MDL number MFCD00069285

  •  PubChem Substance ID 24895056

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience, Enzyme Inhibitors, Enzyme Inhibitors by Enzyme,
InChI Key   GUGJQAFPKZZOIV-UHFFFAOYSA-N
assay   ≥98% (HPLC)
form   powder
color   white
solubility   H2O: soluble10 mM
  ethanol: soluble10 mg/mL
storage temp.   −20°C

Description

Biochem/physiol Actions

Shown to be an intracellular calcium antagonist. Reported to induce neurite formation.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?