• USA Home
  • G1285 - Glucose Regulating Protein 78 from hamster

EMAIL THIS PAGE TO A FRIEND
G1285 Sigma

Glucose Regulating Protein 78 from hamster

recombinant, expressed in E. coli, buffered aqueous solution

Synonym: GRP 78

Purchase

Properties

Related Categories Auxiliary Proteins, Biochemicals and Reagents, Other, Proteins and Derivatives
recombinant   expressed in E. coli
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −70°C

Description

Packaging

Package size based on protein content

Physical form

Solution in 37 mM Tris-HCl, pH 7.5, containing 37 mM NaCl

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?