EMAIL THIS PAGE TO A FRIEND
G130 Sigma

GR 94800

>96%, powder

  • CAS Number 141636-65-9

  • Empirical Formula (Hill Notation) C49H61N9O8

  • Molecular Weight 904.06

  •  MDL number MFCD00214618

Purchase

Properties

Related Categories Antagonist, Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Peptides and Proteins,
InChI Key   CWEISCOCKRHHOU-UJYVDGJVSA-N
assay   >96%
form   powder
color   white
solubility   methanol: soluble1.3 mg/mL
  DMSO: soluble
  H2O: insoluble
storage temp.   −20°C
Gene Information   human ... TACR2(6865)
mouse ... TACR2(21337)
rat ... TACR2(25007)

Description

Amino Acid Sequence

Phenyl-CO-Ala-Ala-Trp-Phe-Pro-Pro-Nle-NH2

Biochem/physiol Actions

Selective NK-2 tachykinin receptor antagonist.

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?