• USA Home
  • G136 - Sp-Guanosine 3′,5′-cyclic monophosphorothioate triethylammonium salt

EMAIL THIS PAGE TO A FRIEND
G136 Sigma

Sp-Guanosine 3′,5′-cyclic monophosphorothioate triethylammonium salt

solid

Synonym: Sp-Cyclic 3′5′-hydrogen phosphorothioate guanosine triethylammonium salt, Sp-cGMPS triethylammonium salt

Purchase

Description

Biochem/physiol Actions

Sp-cGMPS is a metabolically resistant activator of cGMP-dependent protein kinases.

Caution

Hygroscopic

Other Notes

Sp-Diastereomer of guanosine 3′,5′-cyclic monophosphorothioate.

Price and Availability


Better Together

Discover your Target

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?