Synonym: Anti-mGluR1
MDL number MFCD01633636
Popular Documents: Datasheet (PDF)
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Neurotransmission, Antibodies to Glutamate Receptors, |
| species reactivity | human |
| application(s) | immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10 μg/mL using human brain, neurons |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | buffered aqueous solution |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information |
human ... GRM1(2911) mouse ... Grm1(14816) rat ... Grm1(24414) |
| availability | not available in Japan |
synthetic peptide corresponding to the C-terminal of human metabotropic glutamate receptor 1, conjugated to KLH. The immunizing peptide has 88% homology with the rat and mouse gene.
Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Customers Also Viewed
affinity isolated antibody, buffered aqueous glycerol solution

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?