• USA Home
  • G1500 - α-D-Galactose 1-phosphate barium salt

EMAIL THIS PAGE TO A FRIEND
G1500 Sigma

α-D-Galactose 1-phosphate barium salt

Type III, 98-100%

Synonym: α-D-Galactopyranosyl phosphate

  • CAS Number 103213-39-4

  • Linear Formula C6H11O9PBa

  • Molecular Weight 395.45

  •  PubChem Substance ID 329799843

Purchase

Properties

InChI Key   GZVQTLGNBGFOAI-UHFFFAOYSA-N
type   Type III
assay   98-100%
impurities   glucose 1-phosphate., essentially free
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

G0380 α-D-Galactose 1-phosphate dipotassium salt pentahydrate, Type II, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?