• USA Home
  • G1502 - Gly-His hydrochloride hydrate

EMAIL THIS PAGE TO A FRIEND
G1502 Sigma

Gly-His hydrochloride hydrate

Synonym: Glycyl-L-histidine hydrochloride hydrate

  • CAS Number 3486-76-8

  • Empirical Formula (Hill Notation) C8H12N4O3 · HCl · xH2O

  • Molecular Weight 248.67 (anhydrous basis)

  •  Beilstein Registry Number 3743779

  •  MDL number MFCD00167964

  •  PubChem Substance ID 329799844

Purchase

Properties

InChI Key   VGVLXWZHJBRECR-ILKKLZGPSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

G1627 Gly-His

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?