• USA Home
  • G1538 - Gelsolin from human plasma

EMAIL THIS PAGE TO A FRIEND
G1538 Sigma

Gelsolin from human plasma

~90% (SDS-PAGE), lyophilized powder, 100-300 units/mg protein

Purchase

Properties

Related Categories Actin/Actin-binding Proteins, Application Index, Biochemicals and Reagents, Cancer Research, Cell Biology,
assay   ~90% (SDS-PAGE)
form   lyophilized powder
composition   Protein, ~10% Lowry-TCA
solubility   H2O: soluble0.50 - 0.60 mg/mL, clear to slightly hazy
  H2O: soluble0.50 - 0.60 mg/mL, clear to slightly hazy
storage temp.   −20°C
Gene Information   human ... GSN(2934)

Description

Biochem/physiol Actions

Gelsolin is an actin-severing protein found in mammalian cells and blood plasma. Exhibits tumor suppressor activity in several cancer cell lines and in nude mice.

Gelsolin is an actin-binding protein that regulates the assembly and disassembly of actin filaments.

Packaging

Package size based on protein content

Unit Definition

One unit will reduce viscosity difference between actin solution and buffer by 50% in a 1 mL mixture containing 1-2 mg F-actin, 0.15 M KCl, 20 mM Tris, pH 7.6, 0.2 mM CaCl2, 0.2 mM ATP and 1 mM DTT at 28°C.

Physical form

Lyophilized powder containing Tris buffer salts, pH 7.6, NaCl, DTT, and EGTA

Application

Plasma gelsolin levels may be used to identify patients at risk for increased morbidity.

General description

Human gelsolin (also known as brevin) is an actin severing protein found in mammalian cells and blood plasma. Gelsolins consist of a 730 amino acid core structure which contains six repeated domains. These six domains make up three distinct actin binding sites. The plasma enzyme contains an N-terminal extension of 25 amino acids that raises its molecular weight from 90 kD to 93 kD.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?