• USA Home
  • G1788 - Gly-Gly-7-amido-4-methylcoumarin

EMAIL THIS PAGE TO A FRIEND
G1788 Sigma

Gly-Gly-7-amido-4-methylcoumarin

≥97% (TLC)

  • CAS Number 191723-65-6

  • Empirical Formula (Hill Notation) C14H15N3O4

  • Molecular Weight 289.29

  •  MDL number MFCD00152090

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   ZRGZRZYKKHAMID-UHFFFAOYSA-N
assay   ≥97% (TLC)
composition   Peptide content, ~65%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?