EMAIL THIS PAGE TO A FRIEND
G1792 Sigma

GITR Ligand human

>97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder

Synonym: GITRL, Glucocorticoid-induced TNF ligand

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones, TNF Receptors,
recombinant   expressed in baculovirus infected Sf21 cells
assay   >97% (SDS-PAGE)
form   lyophilized powder
mol wt   calculated mol wt 15 kDa
  mol wt 14-16 kDa by SDS-PAGE (triplet)
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   human ... TNFSF18(8995)

Description

Analysis Note

The biological activity is measured by its ability to bind human GITR.

Other Notes

N-terminal histidine-tagged extracellular domain.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 1.25 mg bovine serum albumin.

Biochem/physiol Actions

Type II transmembrane protein in the TNF superfamily. Appears to act in conjunction with the GITR, to protect T cells against activation induced cell death.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?