EMAIL THIS PAGE TO A FRIEND
G2019 Sigma

β-D-Glucose-Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Carbohydrate Matrices, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
matrix   cross-linked 4% beaded agarose
matrix activation   epoxy
matrix attachment   hydroxyl
matrix spacer   12 atoms
capacity   10-20 mg/mL binding capacity (Concanavalin A)
  2-4 mg/mL binding capacity (lectin from Vicia faba)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Application

α-D-Glucose agarose is an agarose conjugate in saline suspension used in affinity chromatography, protein chromatography and carbohydrate matrices. α-D-Glucose -garose has been used in studies to inform pathogenesis of protozoal infections.

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?