• USA Home
  • G2250 - Gangliosides Purified from bovine brain

EMAIL THIS PAGE TO A FRIEND
G2250 Sigma

Gangliosides Purified from bovine brain

Type II

Purchase

Properties

type   Type II
impurities   ≥11% N-acetylneuraminic acid
storage temp.   −20°C

Description

General description

Purified gangliosides (a family of glycosphingolipids) isolated from bovine brain

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?