• USA Home
  • G2400 - Gaskets for vertical standard gels, 16.5 cm wide
EMAIL THIS PAGE TO A FRIEND
G2400 Sigma
Gaskets for vertical standard gels, 16.5 cm wide
size 0.75 mm × 14.5 cm, color-coded yellow
Purchase
Description
Features and Benefits
Unique silicone rubber gasket provides continuous seal around sides and bottom of glass plates, eliminating the need for a bottom spacer. Order gasket thickness to match comb and spacers. Gaskets are color-coded by thickness for easy identification. Length given is height of assembly; actual gasket length is 2 × h + w.
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?