EMAIL THIS PAGE TO A FRIEND
G2627 Sigma

Gly-DL-Phe

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   JBCLFWXMTIKCCB-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?