• USA Home
  • G2653 - Globulin, corticosteroid-binding from human plasma

EMAIL THIS PAGE TO A FRIEND
G2653 Sigma

Globulin, corticosteroid-binding from human plasma

lyophilized powder

Synonym: CBG, Transcortin

Purchase

Description

Biochem/physiol Actions

A 56 kDa glycosylated β-globulin present in serum that regulates the concentration of glucocorticoids and progesterone.

Packaging

Package size based on protein content

Physical form

Lyophilized from 1.25 mM sodium phosphate, pH 7.4, containing 0.05% sodium azide

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?