EMAIL THIS PAGE TO A FRIEND
G2753 Sigma

Glycine hydroxamate

  • CAS Number 5349-80-4

  • Empirical Formula (Hill Notation) C2H6N2O2

  • Molecular Weight 90.08

  •  MDL number MFCD00243727

  •  PubChem Substance ID 24895101

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   VDBCTDQYMZSQFQ-UHFFFAOYSA-N
assay   ≥98%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?