EMAIL THIS PAGE TO A FRIEND
G2771 Sigma

Glass, arylamine

use tested

Purchase

Description

Application

Useful for C-terminal coupling of proteins and peptides.

Preparation Note

Prepared from 75-100 Å aminopropyl controlled pore glass (200-400 mesh) and p-nitrophenylacetic acid, followed by reduction.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?