• USA Home
  • G2884 - L-Glutamine–Agarose

EMAIL THIS PAGE TO A FRIEND
G2884 Sigma

L-Glutamine–Agarose

saline suspension

Purchase

Properties

form   saline suspension
extent of labeling   5-10 μmol per mL
matrix   4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   amino
matrix spacer   1 atom
storage temp.   2-8°C

Description

Physical form

Suspension in 2.0 M NaCl containing 0.02% thimerosal

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?