• USA Home
  • G2885 - γ-Globulins from rat

EMAIL THIS PAGE TO A FRIEND
G2885 Sigma

γ-Globulins from rat

~98% (agarose gel electrophoresis)

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Enzymes, Inhibitors, and Substrates, Globulins, Plasma & Blood Proteins,
assay   ~98% (agarose gel electrophoresis)
impurities   2-4% NaCl
storage temp.   −20°C
Gene Information   rat ... Obp3(259247)

Description

Preparation Note

Prepared from Cohn Fraction II, III

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?