• USA Home
  • G2894 - Anti-Growth Hormone antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
G2894 Sigma

Anti-Growth Hormone antibody produced in rabbit

whole antiserum, lyophilized powder

Synonym: Anti-hGH

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Neurotransmission, Antibodies to Hormones,
species reactivity   human
application(s)   radioimmunoassay: suitable
clone   polyclonal
antibody form   whole antiserum
form   lyophilized powder
contains   0.02% sodium azide
storage temp.   2-8°C
Gene Information   human ... GH1(2688)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

hGH

Specificity

There is no cross-reactivity with glucagon, HCG, TSH, insulin, LH, or prolactin.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Reconstitution

Reconstitute with 1 mL buffer: 0.01 M phosphate, pH 7.4, 0.15 M sodium chloride, 0.1% bovine serum albumin and 15 mM sodium azide. Store the reconstituted stock at −20 °C. Working dilution (1:10) to be discarded if unused within 12 hours.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?