• USA Home
  • G3141 - β-D-Galactopyranosylphenyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
G3141 Sigma

β-D-Galactopyranosylphenyl isothiocyanate

Synonym: Isothiocyanatophenyl β-D-galactopyranoside

  • CAS Number 20721-62-4

  • Empirical Formula (Hill Notation) C13H15NO6S

  • Molecular Weight 313.33

  •  MDL number MFCD00133128

  •  PubChem Substance ID 24895115

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates G, Monosaccharide More...
InChI Key   RWANFUZQWINQBY-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Application

Suitable for the preparation of neoglycoproteins.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?