• USA Home
  • G3521 - L-Glutamic acid amide

EMAIL THIS PAGE TO A FRIEND
G3521 Sigma

L-Glutamic acid amide

Synonym: L-Isoglutamine

  • CAS Number 636-65-7

  • Empirical Formula (Hill Notation) C5H10N2O3

  • Molecular Weight 146.14

  •  MDL number MFCD00135661

  •  PubChem Substance ID 24895129

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   AULUINXDLCVMOX-YFKPBYRVSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

L-Glutamic acid amide is a component of molecules such as the bacterial cell wall muramyl dipeptides and its analogues such as N-Acetylmuramyl-L-alanyl-L-isoglutamine.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?