• USA Home
  • G3526 - L-Glutamine Synthetase from Bacillus stearothermophilus

EMAIL THIS PAGE TO A FRIEND
G3526 Sigma

L-Glutamine Synthetase from Bacillus stearothermophilus

Purchase

Properties

storage temp.   2-8°C

Description

Unit Definition

One unit will convert 1.0 μmole of L-glutamate to L-glutamine in 15 min at pH 7.1 at 37°C.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?