• USA Home
  • G3894 - γ-D-Glutamylaminomethylsulfonic acid

EMAIL THIS PAGE TO A FRIEND
G3894 Sigma

γ-D-Glutamylaminomethylsulfonic acid

Synonym: GAMS

  • CAS Number 90237-02-8

  • Empirical Formula (Hill Notation) C6H12N2O6S

  • Molecular Weight 240.23

  •  PubChem Substance ID 24895142

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   HCMBCMVOCRJSME-SCSAIBSYSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

Gamma-D-Glutamylaminomethylsulfonic acid (GAMS) is used as a selective antagonist of AMPA/kainate glutamatergic receptors and as an anticonvulsant.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?