EMAIL THIS PAGE TO A FRIEND
G3910 Sigma

Glass, carboxymethyl

120-200 mesh, average diameter 80 Å

Synonym: Carboxymethyl-CPG

Purchase

Properties

Related Categories Cation Exchange Media, Ion Exchange Chromatography, Molecular Biology, Protein Chromatography, Proteomics More...
extent of labeling   ~250 μmol carboxymethyl per g
particle size   120-200 mesh
pore size   80 Å average diameter

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?