• USA Home
  • G4165 - Anti-gp130 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
G4165 Sigma

Anti-gp130 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous glycerol solution

Purchase

Description

Immunogen

recombinant GST-gp130 containing the cytoplasmic domain of human gp130.

Physical form

Solution in 0.07 M Tris-glycine, pH 7.4, and 0.105 M NaCl containing 30% glycerol and 0.035% sodium azide.

Preparation Note

Purified using protein A.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?