• USA Home
  • G4283 - Anti-GPR15, N-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
G4283 Sigma

Anti-GPR15, N-Terminal antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-BOB

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Aids and Viral Research, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood,
species reactivity   human, mouse, rat
application(s)   western blot: 1:500-1:2,000 using a human Jurkat (human T cell leukemic), A549 (human lung alveolar epithelial) cell or human heart tissue lysates
clone   polyclonal
concentration   ~0.5 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 50 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... GPR15(2838)
mouse ... Gpr15(71223)
rat ... Gpr15(288181)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the N-terminal and amino acids 13-28 of human GPR15 (BOB). This sequence differs from African green monkey and pig-tailed macaque by one amino acid.

Application

Anti-GPR15, N-Terminal antibody produced in rabbit is suitable for immunoblotting at a working dilution of 1:500-1:2000 using human heart tissue lysate, Jurkat (human T cell leukemic) cell lysate, or A549 (human lung alveolar epithelial) cell lysate.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

GRP15 (G protein-coupled receptor 15) is a seven menbrane, G-protein-coupled receptor with sequence similarity to chemokine receptors and to chemokine receptor-like orphan receptors. It is expressed in lymphoid tissues and colon. It functions as a co-receptor for simian immunodeficiency virus (SIV), strains of HIV-2, and M-tropic HIV-1.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

WGK Germany  -
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?