• USA Home
  • G4388 - Glycerol 3-phosphate Oxidase from Streptococcus thermophilus

EMAIL THIS PAGE TO A FRIEND
G4388 Sigma

Glycerol 3-phosphate Oxidase from Streptococcus thermophilus

lyophilized powder, ≥35 units/mg solid, pH 7.0

Synonym: sn-Glycerol 3-phosphate: oxygen 2-oxidoreductase, L-Glycerol 3-phosphate Oxidase

Purchase

Description

Unit Definition

One unit will oxidize 1.0 μmole of L-glycerol 3-phosphate to dihydroxyacetone phosphate with the formation of H2O2 per min at pH 7.0 at 37 °C.

Application

GPO has been used for sensitive metabolite assays of starch and lipid synthesis, pyrophosphate, ATP, ADP, and most glycolytic intermediates in Arabidopsis seeds. GPO is part of the dihydroxyacetone phosphate:glycerol-3-phosphate cycle in the bloodstream form of Trypanosoma brucei.

Biochem/physiol Actions

Glycerol-3-phosphate oxidase catalyzes the conversion of sn-glycerol-3-phosphate to glycerone phosphate and peroxide.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?