• USA Home
  • G4390 - Globulins Cohn fraction II, III porcine

EMAIL THIS PAGE TO A FRIEND
G4390 Sigma

Globulins Cohn fraction II, III porcine

lyophilized powder

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Enzymes, Inhibitors, and Substrates, Serum Proteins, Serum Proteins and Related Enzymes More...
form   lyophilized powder
composition   Protein, ~75% biuret
storage temp.   2-8°C

Description

Other Notes

Primarily γ-globulins

Physical form

Contains NaCl

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?