EMAIL THIS PAGE TO A FRIEND
G4518 Sigma
Aminopropyl–CPG
200-400 mesh, average pore size 170 Å
Purchase
Properties
extent of labeling   125-225 μmol primary amine per g glass
particle size   200-400 mesh
pore size   170 Å average pore size
suitability   suitable for attaching proteins, peptides and ligands (Contains primary amino groups)
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?